Boc-(S)-3-amino-5-phenylpentanoic acid 95%

Supplier: Thermo Scientific

H52180.MD H52180.03
ALFAH52180.MDEA 0 SGD
ALFAH52180.MD ALFAH52180.03
Boc-(S)-3-amino-5-phenylpentanoic acid 95%
Boc-(S)-3-amino-5-phenylpentanoic acid

Purity: 95% CAS No.: 218608-84-5 Molecular Formula: C16H23NO4 Formula Weight: 293.36 MDL No.: MFCD01076264

Formula: C₁₆H₂₃NO₄
MW: 293,37 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01076264
CAS Number: 218608-84-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR