Boc-L-β-homoglutamic acid-d-benzyl ester 95%
Supplier: Thermo Scientific
H52187.MD
H52187.03
ALFAH52187.MDEA
0
SGD
ALFAH52187.MD
ALFAH52187.03
Boc-L-β-homoglutamic acid-d-benzyl ester 95%
Boc-L-beta-homoglutamic acid-d-benzyl ester
Purity: 95%
CAS No.: 218943-30-7
Molecular Formula: C18H25NO6
Formula Weight: 351.40
Beilstein Reference: 8079168
MDL No.: MFCD01862936
Formula:
C₁₈H₂₅NO₆ MW: 351,4 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01862936 CAS Number: 218943-30-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...