4-(6-Methyl-2-benzimidazolyl)benzamidoxime 97%
Supplier: Thermo Scientific
|
Warning
|
H52247.MD
H52247.03
H52247.06
ALFAH52247.MDEA
129
SGD
ALFAH52247.MD
ALFAH52247.03
ALFAH52247.06
4-(6-Methyl-2-benzimidazolyl)benzamidoxime 97%
4-(6-Methyl-2-benzimidazolyl)benzamidoxime
Purity: 97%
Molecular Formula: C15H14N4O
Formula Weight: 266.30
MDL No.: MFCD16658868
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...