2-Chlorothionicotinamide 97%

Supplier: Thermo Scientific
Warning

Synonyms: 2-Chloropyridine-3-thiocarboxamide

H52252.03 H52252.06
ALFAH52252.03EA 87.2 SGD
ALFAH52252.03 ALFAH52252.06
2-Chlorothionicotinamide 97%
2-Chlorothionicotinamide

Purity: 97% Molecular Formula: C6H5ClN2S Formula Weight: 172.63 MDL No.: MFCD16659867

Formula: C₆H₅ClN₂S
MW: 172,63 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16659867
CAS Number: 1240596-59-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR