4-(6-Amidoximo-2-benzimidazolyl)benzamidoxime 97%
Supplier: Thermo Scientific
|
Warning
|
H52261.MD
H52261.03
ALFAH52261.MDEA
215
SGD
ALFAH52261.MD
ALFAH52261.03
4-(6-Amidoximo-2-benzimidazolyl)benzamidoxime 97%
4-(6-Amidoximo-2-benzimidazolyl)benzamidoxime
Purity: 97%
Molecular Formula: C15H14N6O2
Formula Weight: 310.32
MDL No.: MFCD16660854
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...