9-Benzyl-6-chloro-7-iodo-7-deazapurine 95%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
7-Benzyl-4-chloro-5-iodo-7H-pyrrolo[2,3-d]pyrimidine
H52275.03
H52275.06
ALFAH52275.03EA
172
SGD
ALFAH52275.03
ALFAH52275.06
9-Benzyl-6-chloro-7-iodo-7-deazapurine 95%
9-Benzyl-6-chloro-7-iodo-7-deazapurine
Purity: 95%
Molecular Formula: C13H9ClIN3
Formula Weight: 369.59
MDL No.: MFCD16660858
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...