2-Bromo-3-(chloromethyl)pyridine 96%

Supplier: Thermo Scientific
Warning

H52313.MD H52313.03
ALFAH52313.MDEA 0 SGD
ALFAH52313.MD ALFAH52313.03
2-Bromo-3-(chloromethyl)pyridine 96%
2-Bromo-3-(chloromethyl)pyridine

Purity: 96% CAS No.: 944906-87-0 Molecular Formula: C6H5BrClN Formula Weight: 206.47 Storage Temperature: 2°C to 8°C Melting Point: 34-36° Beilstein Reference: 15651009 MDL No.: MFCD10697618

Formula: C₆H₅BrClN
MW: 206,47 g/mol
Melting Pt: 34…36 °C
MDL Number: MFCD10697618
CAS Number: 944906-87-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR