4-Carbamoyl-3,5-difluorobenzeneboronic acid pinacol ester 96%

Supplier: Thermo Scientific

Synonyms: 2,6-Difluoro-4-(4,4,5,5-tetramethyl-[1,3,2]dioxaborolan-2-yl)benzamide

H52316.MD H52316.03
ALFAH52316.MDEA 199 SGD
ALFAH52316.MD ALFAH52316.03
4-Carbamoyl-3,5-difluorobenzeneboronic acid pinacol ester 96%
4-Carbamoyl-3,5-difluorobenzeneboronic acid pinacol ester

Purity: 96% Molecular Formula: C13H16BF2NO3 Formula Weight: 283.08 MDL No.: MFCD17215212

Formula: C₁₃H₁₆BF₂NO₃
MW: 283,08 g/mol
Storage Temperature: Ambient
MDL Number: MFCD17215212
CAS Number: 957345-69-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR