2-(Piperazin-1-yl)thiazole hydrochloride ≥97%
Supplier: Thermo Scientific
|
Warning
|
H52368.MD
H52368.03
H52368.06
ALFAH52368.MDEA
134
SGD
ALFAH52368.MD
ALFAH52368.03
ALFAH52368.06
2-(Piperazin-1-yl)thiazole hydrochloride ≥97%
2-(Piperazin-1-yl)thiazole hydrochloride
Purity: 97+%
CAS No.: 209733-13-1
Molecular Formula: C7H11N3S.HCl
Formula Weight: 205.71
MDL No.: MFCD08752687
Formula:
C₇H₁₂ClN₃S MW: 205,71 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD08752687 CAS Number: 209733-13-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...