Methyl-3-(2-bromoethoxy)benzenecarboxylate ≥97%
Supplier: Thermo Scientific
|
Warning
|
H52405.MD
H52405.03
ALFAH52405.MDEA
0
SGD
ALFAH52405.MD
ALFAH52405.03
Methyl-3-(2-bromoethoxy)benzenecarboxylate ≥97%
Methyl-3-(2-bromoethoxy)benzenecarboxylate
Purity: 97+%
CAS No.: 59516-96-0
Molecular Formula: C10H11BrO3
Formula Weight: 259.10
MDL No.: MFCD06204041
Formula:
C₁₀H₁₁BrO₃ MW: 259,11 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD06204041 CAS Number: 59516-96-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...