(S)-N-Boc-3-piperidinemethanol 97%

Supplier: Thermo Scientific
Warning

Synonyms: (s)-n-(tert-butoxycarbonyl)-3-hydroxymethylpiperidine, (s)-tert-butyl 3-(hydroxymethyl)tetrahydro-1(2h)-pyridinecarboxylate, (S)-1-BOC-3-(Hydroxymethyl)piperidine, tert-Butyl (S)-3-(hydroxymethyl)piperidine-1-carboxylate

H52435.03 H52435.06
ALFAH52435.03EA 0 SGD
ALFAH52435.03 ALFAH52435.06
(S)-N-Boc-3-piperidinemethanol 97%
(S)-N-Boc-3-piperidinemethanol

Purity: 97% CAS No.: 140695-84-7 Molecular Formula: C11H21NO3 Formula Weight: 215.29 MDL No.: MFCD02683203

Formula: C₁₁H₂₁NO₃
MW: 215,293 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02683203
CAS Number: 140695-84-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR