3-Formyl-N-(tert-butoxycarbonyl)piperidine 97%

Supplier: Thermo Scientific
Warning

Synonyms: Piperidine-3-carboxaldehyde, N-BOC protected, tert-Butyl 3-formylpiperidine-1-carboxylate

H52571.MD H52571.03 H52571.06
ALFAH52571.MDEA 91.3 SGD
ALFAH52571.MD ALFAH52571.03 ALFAH52571.06
3-Formyl-N-(tert-butoxycarbonyl)piperidine 97%
3-Formyl-N-(tert-butoxycarbonyl)piperidine

Purity: 97% CAS No.: 118156-93-7 Molecular Formula: C11H19NO3 Formula Weight: 213.28 MDL No.: MFCD02179020

Formula: C₁₁H₁₉NO₃
MW: 213,28 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02179020
CAS Number: 118156-93-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR