a-Tosyl(4-fluorobenzyl)isocyanide 97%

Supplier: Thermo Scientific
Danger

Synonyms: (4-Fluorophenyl)(isocyano)methyl 4-methylphenyl sulphone, a-Tosyl-(4-fluorobenzyl) isocyanide

H52580.MD H52580.03
ALFAH52580.MDEA 0 SGD
ALFAH52580.MD ALFAH52580.03
a-Tosyl(4-fluorobenzyl)isocyanide 97%
a-Tosyl(4-fluorobenzyl)isocyanide

Purity: 97% CAS No.: 165806-95-1 Molecular Formula: C15H12FNO2S Formula Weight: 289.32 MDL No.: MFCD03787932

Formula: C₁₅H₁₂FNO₂S
MW: 289,32 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03787932
CAS Number: 165806-95-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR