1-(tert-Butyldimethylsilyl)indole-5-boronic acid 97%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
(1-(tert-Butyldimethylsilyl)-1H-indol-5-yl)boronic acid
H52624.MD
H52624.03
ALFAH52624.MDEA
100
SGD
ALFAH52624.MD
ALFAH52624.03
1-(tert-Butyldimethylsilyl)indole-5-boronic acid 97%
1-(tert-Butyldimethylsilyl)indole-5-boronic acid
Purity: 97%
CAS No.: 913835-68-4
Molecular Formula: C14H22BNO2Si
Formula Weight: 275.23
Storage Temperature: -30°C to -10°C
Melting Point: 248-250°
MDL No.: MFCD03095156
Formula:
C₁₄H₂₂BNO₂Si MW: 275,23 g/mol Melting Pt: 248…250 °C |
MDL Number:
MFCD03095156 CAS Number: 913835-68-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...