5-Fluoro-2-hydroxybenzeneboronic acid 96%

Supplier: Thermo Scientific
Warning

Synonyms: 5-Fluoro-2-hydroxyphenylboronic acid, (5-Fluoro-2-hydroxyphenyl)boronic acid, 5-Fluoro-2-hydroxybenzeneboronic acid

H52716.MD H52716.03
ALFAH52716.MDEA 78 SGD
ALFAH52716.MD ALFAH52716.03
5-Fluoro-2-hydroxybenzeneboronic acid 96%
5-Fluoro-2-hydroxybenzeneboronic acid

Purity: 96% CAS No.: 259209-20-6 Molecular Formula: C6H6BFO3 Formula Weight: 155.92 Physical Form: Solid Appearance: Gray Melting Point: 160-166° MDL No.: MFCD03095357

Formula: C₆H₆BFO₃
MW: 155,92 g/mol
Melting Pt: 160…166 °C
Storage Temperature: Ambient
MDL Number: MFCD03095357
CAS Number: 259209-20-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR