4-Dimethoxymethyl-2-(trifluoromethyl)pyrimidine 97%

Supplier: Thermo Scientific
Warning

H52736.MD H52736.03
ALFAH52736.MDEA 242 SGD
ALFAH52736.MD ALFAH52736.03
4-Dimethoxymethyl-2-(trifluoromethyl)pyrimidine 97%
4-Dimethoxymethyl-2-(trifluoromethyl)pyrimidine

Purity: 97% CAS No.: 878760-47-5 Molecular Formula: C8H9F3N2O2 Formula Weight: 222.17 MDL No.: MFCD05662688

Formula: C₈H₉F₃N₂O₂
MW: 222,17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD05662688
CAS Number: 878760-47-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR