(S)-1,3-Dibenzylpiperazine 97%

Supplier: Thermo Scientific
Warning

H52739.MD H52739.03
ALFAH52739.MDEA 0 SGD
ALFAH52739.MD ALFAH52739.03
(S)-1,3-Dibenzylpiperazine 97%
(S)-1,3-Dibenzylpiperazine

Purity: 97% CAS No.: 204327-96-8 Molecular Formula: C18H22N2 Formula Weight: 266.39 MDL No.: MFCD03787922

Formula: C₁₈H₂₂N₂
MW: 266,39 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03787922
CAS Number: 204327-96-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR