(S)-3-Amino-1-benzylpiperidine 97%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
(S)-1-Benzylpiperidin-3-amine
H52772.03
H52772.06
ALFAH52772.03EA
131
SGD
ALFAH52772.03
ALFAH52772.06
(S)-3-Amino-1-benzylpiperidine 97%
(S)-3-Amino-1-benzylpiperidine
Purity: 97%
CAS No.: 168466-85-1
Molecular Formula: C12H18N2
Formula Weight: 190.29
MDL No.: MFCD03093375
Formula:
C₁₂H₁₈N₂ MW: 190,29 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03093375 CAS Number: 168466-85-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...