Boc-D-α-methylserine 97%
Supplier: Thermo Scientific
H52787.MD
H52787.03
ALFAH52787.MDEA
410
SGD
ALFAH52787.MD
ALFAH52787.03
Boc-D-α-methylserine 97%
Boc-D-alpha-methylserine
Purity: 97%
CAS No.: 84311-18-2
Molecular Formula: C9H17NO5
Formula Weight: 219.24
MDL No.: MFCD02682596
Formula:
C₉H₁₇NO₅ MW: 219,24 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD02682596 CAS Number: 84311-18-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...