O-Ethoxy-L-serine hydrochloride 97%
Supplier: Thermo Scientific
H52790.MD
H52790.03
ALFAH52790.MDEA
373
SGD
ALFAH52790.MD
ALFAH52790.03
O-Ethoxy-L-serine hydrochloride 97%
O-Ethoxy-L-serine hydrochloride
Purity: 97%
CAS No.: 4775-82-0
Molecular Formula: C5H11NO3.HCl
Formula Weight: 169.61
MDL No.: MFCD06797557
Formula:
C₅H₁₁NO₃·HCl MW: 169,61 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD06797557 CAS Number: 4775-82-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...