(R)-2-Hydroxy-4-phenylbutyric acid 97%
Supplier: Thermo Scientific
H52804.03
H52804.06
ALFAH52804.03EA
103
SGD
ALFAH52804.03
ALFAH52804.06
(R)-2-Hydroxy-4-phenylbutyric acid 97%
(R)-2-Hydroxy-4-phenylbutyric acid
Purity: 97%
CAS No.: 29678-81-7
Molecular Formula: C10H12O3
Formula Weight: 180.20
Melting Point: 114-117°
Beilstein Reference: 3201296
Optical Rotation: -9.5° (c=2.8 in ethanol)
MDL No.: MFCD00192219
Formula:
C₁₀H₁₂O₃ MW: 180,20 g/mol Melting Pt: 114…117 °C Storage Temperature: Ambient |
MDL Number:
MFCD00192219 CAS Number: 29678-81-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...