3-(Methylsulphonylamino)phenylboronic acid 97%

Supplier: Thermo Scientific
Warning

Synonyms: 3-Methylsulphonylaminophenylboronic acid, 3-(Methylsulphonylamino)phenylboronic Acid

H52895.03 H52895.06
ALFAH52895.03EA 115 SGD
ALFAH52895.03 ALFAH52895.06
3-(Methylsulphonylamino)phenylboronic acid 97%
3-(Methylsulphonylamino)phenylboronic acid

Purity: 97% CAS No.: 148355-75-3 Molecular Formula: C7H10BNO4S Formula Weight: 215.03 Melting Point: 90-96° MDL No.: MFCD02179478

Formula: C₇H₁₀BNO₄S
MW: 215,04 g/mol
Melting Pt: 90…96 °C
Storage Temperature: Ambient
MDL Number: MFCD02179478
CAS Number: 148355-75-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR