4-(Methoxycarbonylamino)benzeneboronic acid 97%

Supplier: Thermo Scientific
Warning

Synonyms: (4-((Methoxycarbonyl)amino)phenyl)boronic acid

H53027.MD H53027.03
ALFAH53027.MDEA 0 SGD
ALFAH53027.MD ALFAH53027.03
4-(Methoxycarbonylamino)benzeneboronic acid 97%
4-(Methoxycarbonylamino)benzeneboronic acid

Purity: 97% CAS No.: 850567-96-3 Molecular Formula: C8H10BNO4 Formula Weight: 194.98 Melting Point: 234-236° MDL No.: MFCD03788425

Formula: C₈H₁₀BNO₄
MW: 194,98 g/mol
Melting Pt: 234…236 °C
Storage Temperature: Ambient
MDL Number: MFCD03788425
CAS Number: 850567-96-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR