4-(5,6-Dihydro-4H-1,3-oxazin-2-yl)benzeneboronic acid 96%

Supplier: Thermo Scientific
Warning

H53062.MD H53062.03
ALFAH53062.MDEA 0 SGD
ALFAH53062.MD ALFAH53062.03
4-(5,6-Dihydro-4H-1,3-oxazin-2-yl)benzeneboronic acid 96%
4-(5,6-Dihydro-4H-1,3-oxazin-2-yl)benzeneboronic acid

Purity: 96% CAS No.: 850568-68-2 Molecular Formula: C10H12BNO3 Formula Weight: 205.02 Melting Point: 180-182° MDL No.: MFCD06659833

Formula: C₁₀H₁₂BNO₃
MW: 205,02 g/mol
Melting Pt: 180…182 °C
Storage Temperature: Ambient
MDL Number: MFCD06659833
CAS Number: 850568-68-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR