2-Aminomethyl-5-fluorobenzeneboronic acid hydrochloride 95%

Supplier: Thermo Scientific
Warning

H53097.MD H53097.03
ALFAH53097.MDEA 174 SGD
ALFAH53097.MD ALFAH53097.03
2-Aminomethyl-5-fluorobenzeneboronic acid hydrochloride 95%
2-Aminomethyl-5-fluorobenzeneboronic acid hydrochloride

Purity: 95% CAS No.: 850568-03-5 Molecular Formula: C7H9BFNO2.HCl Formula Weight: 205.42 Melting Point: 152-158° MDL No.: MFCD04115666

Formula: C₇H₉BFNO₂·HCl
MW: 205,42 g/mol
Melting Pt: 152…158 °C
Storage Temperature: Ambient
MDL Number: MFCD04115666
CAS Number: 850568-03-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR