4-(2-Bromoethoxy)benzeneboronic acid pinacol ester 97%
Supplier: Thermo Scientific
H53246.MD
H53246.03
ALFAH53246.MDEA
0
SGD
ALFAH53246.MD
ALFAH53246.03
4-(2-Bromoethoxy)benzeneboronic acid pinacol ester 97%
4-(2-Bromoethoxy)benzeneboronic acid pinacol ester
Purity: 98%
CAS No.: 913836-27-8
Molecular Formula: C14H20BBrO3
Formula Weight: 327.03
Melting Point: 90-92°
MDL No.: MFCD09027285
Formula:
C₁₄H₂₀BBrO₃ Melting Pt: 90…92 °C Storage Temperature: Ambient |
MDL Number:
MFCD09027285 CAS Number: 913836-27-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...