2,9-Dimethyl-5-decyne 96%
Supplier: Thermo Scientific
|
Warning
|
H53414.03
H53414.06
ALFAH53414.03EA
136
SGD
ALFAH53414.03
ALFAH53414.06
2,9-Dimethyl-5-decyne 96%
2,9-Dimethyl-5-decyne
Purity: 96%
CAS No.: 19550-56-2
Molecular Formula: C12H22
Formula Weight: 166.31
Boiling Point: 92°/19mm
MDL No.: MFCD00041611
Formula:
C₁₂H₂₂ MW: 166,31 g/mol Boiling Pt: 92 °C (19 mmHg) Density: 0,77 Storage Temperature: Ambient |
MDL Number:
MFCD00041611 CAS Number: 19550-56-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...