2-Methylpropane-1-sulphonyl chloride 97%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
Isobutanesulphonyl chloride, 1-Propanesulphonylchloride, 2-methyl-
Purity: 97%
CAS No.: 35432-36-1
Molecular Formula: C4H9ClO2S
Formula Weight: 156.62
Flash Point: 88°(190°F)
Physical Form: Liquid
Density: 1.20
Refractive Index: 1.4540
Sensitivity: Moisture Sensitive
Appearance: Colourless
Beilstein Reference: 1748728
UN No.: UN3265
MDL No.: MFCD07369800
Formula:
C₄H₉ClO₂S MW: 156,624 g/mol Density: 1,20 Flash Pt: 88 °C (190 °F) Storage Temperature: Ambient |
MDL Number:
MFCD07369800 CAS Number: 35432-36-1 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...