Thiamine nitrate
Supplier: Thermo Scientific
Synonyms:
Vitamin B1 Nitrate
J60014.09
J60014.14
ALFAJ60014.09EA
156
SGD
ALFAJ60014.09
ALFAJ60014.14
Thiamine nitrate
Thiamine nitrate
CAS No.: 532-43-4
Molecular Formula: C12H17N5O4S
Formula Weight: 327.36
Physical Form: Powder
Appearance: White
Melting Point: 190°
MDL No.: MFCD00036330
Formula:
C₁₂H₁₇N₅O₄S MW: 327,36 g/mol Melting Pt: 190 °C Storage Temperature: Ambient |
MDL Number:
MFCD00036330 CAS Number: 532-43-4 EINECS: 208-537-4 Merck Index: 14,9295 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...