Myricetin 98%

Supplier: Thermo Scientific

Synonyms: 3,3',4',5,5',7-Hexahydroxyflavone, 3,5,7-Trihydroxy-2-(3,4,5-trihydroxyphenyl)-4H-chromen-4-one

J60450.MA J60450.MB J60450.MF
ALFAJ60450.MAEA 239 SGD
ALFAJ60450.MA ALFAJ60450.MB ALFAJ60450.MF
Myricetin 98%
Myricetin

Purity: 98% CAS No.: 529-44-2 Molecular Formula: C15H10O8 Formula Weight: 318.24 Storage Temperature: -30°C to -10°C Physical Form: Crystalline solid Appearance: Yellowish brown Melting Point: 300° Solubility: Soluble in DMSO or DMF at approximately 10mg/ml. Also soluble in ethanol at 1mg/ml Beilstein Reference: 332331 MDL No.: MFCD00006827


Applications: Common flavanol in plants that inhibits phenolsulfotransferase. Strongly inhibits yeast alpha-glucosidase

Formula: C₁₅H₁₀O₈
MW: 318,24 g/mol
Melting Pt: >300 °C
Storage Temperature: Freezer
MDL Number: MFCD00006827
CAS Number: 529-44-2
EINECS: 208-463-2
Merck Index: 14,6332

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR