India ink 0,2%
Supplier: Thermo Scientific
J61007.AP
J61007.K2
ALFAJ61007.APEA
353
SGD
ALFAJ61007.AP
ALFAJ61007.K2
India ink 0,2%
India ink
CAS No.: 8046-52-4
Physical Form: Liquid
Appearance: Black
Odour: Odourless
MDL No.: MFCD00147244
Applications: For negative staining of microorganisms
Storage Temperature:
Ambient |
MDL Number:
MFCD00147244 CAS Number: 8046-52-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...