Ethyl-3-ethyl-5-methylisoxazole-4-carboxylate 98%

Supplier: Thermo Fisher Scientific

L00526.06 L00526.14
ALFAL00526.06EA 0 SGD
ALFAL00526.06 ALFAL00526.14
Ethyl-3-ethyl-5-methylisoxazole-4-carboxylate 98%
Ethyl-3-ethyl-5-methylisoxazole-4-carboxylate

Purity: 98% CAS No.: 53064-41-8 Molecular Formula: C9H13NO3 Formula Weight: 183.21 Flash Point: 103°(217°F) Refractive Index: 1.4630 Boiling Point: 71-72°/0.5mm Beilstein Reference: 1073370 MDL No.: MFCD00014094

Formula: C₉H₁₃NO₃
Boiling Pt: 71…72 °C (0,5 mmHg)
Density: 1,07
Flash Pt: 103 °C (217 °F)
Storage Temperature: Ambient
MDL Number: MFCD00014094
CAS Number: 53064-41-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR