4-Methoxybenzyl carbazate 97%

Supplier: Thermo Scientific
Warning

Synonyms: 4-Methoxybenzyl hydrazinecarboxylate

L00541.03 L00541.06
ALFAL00541.03EA 0 SGD
ALFAL00541.03 ALFAL00541.06
4-Methoxybenzyl carbazate 97%
4-Methoxybenzyl carbazate

Purity: 97% CAS No.: 18912-37-3 Molecular Formula: C9H12N2O3 Formula Weight: 196.21 Melting Point: 75-77° Beilstein Reference: 2052164 MDL No.: MFCD00042817

Formula: C₉H₁₂N₂O₃
MW: 196,21 g/mol
Melting Pt: 75…77 °C
Storage Temperature: Ambient
MDL Number: MFCD00042817
CAS Number: 18912-37-3
EINECS: 242-664-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR