Pentafluoropropionaldehyde hydrate 90%, tech.
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
2,2,3,3,3-Pentafluoropropane-1,1-diol
L00810.06
L00810.14
ALFAL00810.06EA
0
SGD
ALFAL00810.06
ALFAL00810.14
Pentafluoropropionaldehyde hydrate 90%, tech.
Pentafluoropropionaldehyde hydrate
Purity: 90%
CAS No.: 422-63-9
Molecular Formula: C3H3F5O2.xH2O
Formula Weight: 166.05(anhy)
Refractive Index: 1.3250
Melting Point: 50-54°
Beilstein Reference: 1705233
MDL No.: MFCD00792902
Formula:
C₃H₃F₅O₂·xH₂O MW: 166,05 g/mol (anhy) Melting Pt: 50…54 °C Storage Temperature: Ambient |
MDL Number:
MFCD00792902 CAS Number: 422-63-9 EINECS: 207-020-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...