1,3-Di-2-thienyl-2-propen-1-one ≥98%
Supplier: Thermo Scientific
L00857.06
L00857.14
ALFAL00857.06EA
91.7
SGD
ALFAL00857.06
ALFAL00857.14
1,3-Di-2-thienyl-2-propen-1-one ≥98%
1,3-Di-2-thienyl-2-propen-1-one
Purity: 98+%
CAS No.: 2309-48-0
Molecular Formula: C11H8OS2
Formula Weight: 220.31
Melting Point: 98-100°
Beilstein Reference: 143294
MDL No.: MFCD00014530
Formula:
C₁₁H₈OS₂ MW: 220,31 g/mol Melting Pt: 98…100 °C Storage Temperature: Ambient |
MDL Number:
MFCD00014530 CAS Number: 2309-48-0 EINECS: 218-993-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...