1-(4-Ethylphenyl)-1H-pyrrole-2,5-dione ≥98%

Supplier: Thermo Scientific
Danger

L01058.04 L01058.09
ALFAL01058.04EA 160 SGD
ALFAL01058.04 ALFAL01058.09
1-(4-Ethylphenyl)-1H-pyrrole-2,5-dione ≥98%
1-(4-Ethylphenyl)-1H-pyrrole-2,5-dione

Purity: 98+% CAS No.: 76620-00-3 Molecular Formula: C12H11NO2 Formula Weight: 201.22 Melting Point: 64-67° Beilstein Reference: 7022079 MDL No.: MFCD00014538

Formula: C₁₂H₁₁NO₂
Melting Pt: 64…67 °C
Storage Temperature: Ambient
MDL Number: MFCD00014538
CAS Number: 76620-00-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR