2-Carboxycinnamic acid predominantly trans 97%
Supplier: Thermo Scientific
|
Warning
|
L01467.06
L01467.14
ALFAL01467.06EA
89.6
SGD
ALFAL01467.06
ALFAL01467.14
2-Carboxycinnamic acid predominantly trans 97%
2-Carboxycinnamic acid
Purity: 97%
CAS No.: 612-40-8
Molecular Formula: C10H8O4
Formula Weight: 192.17
Melting Point: 195-205°
Beilstein Reference: 1870294
MDL No.: MFCD00004380
Formula:
C₁₀H₈O₄ MW: 192,17 g/mol Melting Pt: 195…205 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004380 CAS Number: 612-40-8 EINECS: 210-308-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...