1-(2-Cyanoethyl)piperidine ≥98%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
Piperidinopropionitrile
L01469.30
L01469.18
ALFAL01469.30EA
0
SGD
ALFAL01469.30
ALFAL01469.18
1-(2-Cyanoethyl)piperidine ≥98%
1-(2-Cyanoethyl)piperidine
Purity: 98+%
CAS No.: 3088-41-3
Molecular Formula: C8H14N2
Formula Weight: 138.21
Flash Point: 102°(215°F)
Refractive Index: 1.4700
Boiling Point: 110-111°/16mm
Beilstein Reference: 107858
UN No.: UN3276
MDL No.: MFCD00006511
Formula:
C₈H₁₄N₂ MW: 138,21 g/mol Boiling Pt: 110…111 °C (16 mmHg) Density: 0,933 Flash Pt: 102 °C (215 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00006511 CAS Number: 3088-41-3 EINECS: 221-417-6 UN: 3276 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...