4-Amino-6-hydroxy-2-mercaptopyrimidine hydrate ≥98%

Supplier: Thermo Scientific

Synonyms: 4-Amino-6-hydroxy-2-mercaptopyrimidine monohydrate, 4-Amino-6-hydroxy-2-mercaptopyrimidine Hydrate, 6-Amino-2-thiouracil hydrate, 6-Amino-2-thiouracil Hydrate

L01992.14 L01992.22
ALFAL01992.14EA 73.6 SGD
ALFAL01992.14 ALFAL01992.22
4-Amino-6-hydroxy-2-mercaptopyrimidine hydrate ≥98%
4-Amino-6-hydroxy-2-mercaptopyrimidine hydrate

4-Amino-6-hydroxy-2-mercaptopyrimidine is a novel, potent, and selective A3 adenosine receptor antagonists. It is also used as pharmaceutical intermediates.


Purity: 98+% CAS No.: 65802-56-4 Molecular Formula: C4H5N3OS.H2O Formula Weight: 161.19 (143.17anhy) Melting Point: 300° Beilstein Reference: 5795441 MDL No.: MFCD00150556

Formula: C₄H₅N₃OS·xH₂O (x=1)
MW: 143,17 g/mol (anhy)
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00150556
CAS Number: 65802-56-4
EINECS: 213-722-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR