5,6-Dihydro-6-methyluracil 99%

Supplier: Thermo Scientific

L02292.03 L02292.06
ALFAL02292.03EA 0 SGD
ALFAL02292.03 ALFAL02292.06
5,6-Dihydro-6-methyluracil 99%
5,6-Dihydro-6-methyluracil

Purity: 99% CAS No.: 2434-49-3 Molecular Formula: C5H8N2O2 Formula Weight: 128.13 Melting Point: 218-220° Beilstein Reference: 118196 MDL No.: MFCD00023160

Formula: C₅H₈N₂O₂
MW: 128,13 g/mol
Melting Pt: 218…220 °C
Storage Temperature: Ambient
MDL Number: MFCD00023160
CAS Number: 2434-49-3
EINECS: 219-421-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR