Dimethyl (phthalimidomethyl)phosphonate 97%

Supplier: Thermo Scientific

L02840.04 L02840.09 L02840.18
ALFAL02840.04EA 84.2 SGD
ALFAL02840.04 ALFAL02840.09 ALFAL02840.18
Dimethyl (phthalimidomethyl)phosphonate 97%
Dimethyl (phthalimidomethyl)phosphonate

Purity: 97% CAS No.: 28447-26-9 Molecular Formula: C11H12NO5P Formula Weight: 269.19 Melting Point: 115-120° Beilstein Reference: 1476370 MDL No.: MFCD00015464

Formula: C₁₁H₁₂NO₅P
MW: 269,19 g/mol
Melting Pt: 115…120 °C
Storage Temperature: Ambient
MDL Number: MFCD00015464
CAS Number: 28447-26-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR