1H-Pyrazolo[3,4-d]pyrimidin-4-amine 98%

Supplier: Thermo Scientific
Danger

Synonyms: 1H-Pyrazolo[3,4-d]pyrimidine-4-ylamine 4-Amino-1H-pyrazolo[3,4-d]pyrimidine, 4-Amino-1H-pyrazolo[3,4-d]pyrimidine, 4-Aminopyrazolo[3,4-d]pyrimidine

L03519.03 L03519.06
ALFAL03519.03EA 0 SGD
ALFAL03519.03 ALFAL03519.06
1H-Pyrazolo[3,4-d]pyrimidin-4-amine 98%
1H-Pyrazolo[3,4-d]pyrimidin-4-amine

Purity: 98% CAS No.: 2380-63-4 Molecular Formula: C5H5N5 Formula Weight: 135.13 Odour: Odourless Melting Point: 310° dec. Beilstein Reference: 5824 UN No.: UN2811 MDL No.: MFCD00005688

Formula: C₅H₅N₅
MW: 135,13 g/mol
Melting Pt: >310 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00005688
CAS Number: 2380-63-4
EINECS: 243-691-6
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR