(Benzylthio)acetone 98%

Supplier: Thermo Scientific

L03814.06 L03814.14
ALFAL03814.06EA 0 SGD
ALFAL03814.06 ALFAL03814.14
(Benzylthio)acetone 98%
(Benzylthio)acetone

Purity: 98% CAS No.: 10230-69-0 Molecular Formula: C10H12OS Formula Weight: 180.27 Flash Point: 100°(212°F) Refractive Index: 1.5580 Boiling Point: 155-156°/17mm Beilstein Reference: 1866268 MDL No.: MFCD00026241

Formula: C₁₀H₁₂OS
MW: 180,27 g/mol
Boiling Pt: 155…156 °C (17 mmHg)
Density: 1,090
Flash Pt: >100 °C (212 °F)
Storage Temperature: Ambient
MDL Number: MFCD00026241
CAS Number: 10230-69-0
EINECS: 233-552-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR