4-Amino-6-hydroxypyrimidine 98%
Supplier: Thermo Scientific
|
Warning
|
L03825.03
L03825.06
ALFAL03825.03EA
0
SGD
ALFAL03825.03
ALFAL03825.06
4-Amino-6-hydroxypyrimidine 98%
4-Amino-6-hydroxypyrimidine
Purity: 98%
CAS No.: 1193-22-2
Molecular Formula: C4H5N3O
Formula Weight: 111.10
Melting Point: 262-264°
Beilstein Reference: 112429
MDL No.: MFCD00051502
Formula:
C₄H₅N₃O MW: 111,10 g/mol Melting Pt: 262…264 °C Storage Temperature: Ambient |
MDL Number:
MFCD00051502 CAS Number: 1193-22-2 EINECS: 214-771-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...