Chalcone-α,β-epoxide 98%

Supplier: Thermo Scientific
Warning

Synonyms: Chalcone Oxide, 2-Benzoyl-3-phenyloxirane, 1,3-Diphenyl-2,3-epoxy-1-propanone, 1,3-Diphenyl-2-propen-1-one Oxide

L03874.06 L03874.14
ALFAL03874.06EA 0 SGD
ALFAL03874.06 ALFAL03874.14
Chalcone-α,β-epoxide 98%
Chalcone-alpha,beta-epoxide

Purity: 98% CAS No.: 5411-12-1 Molecular Formula: C15H12O2 Formula Weight: 224.26 Melting Point: 88-90° Beilstein Reference: 164553 MDL No.: MFCD00022336

Formula: C₁₅H₁₂O₂
MW: 224,26 g/mol
Melting Pt: 88…90 °C
Storage Temperature: Ambient
MDL Number: MFCD00022336
CAS Number: 5411-12-1
EINECS: 226-487-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR