N-Methyl-N-trimethylsilylacetamide 98%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
N-Methyl-N-TMS-acetamide
L04101.04
L04101.09
ALFAL04101.04EA
73.2
SGD
ALFAL04101.04
ALFAL04101.09
N-Methyl-N-trimethylsilylacetamide 98%
N-Methyl-N-trimethylsilylacetamide
Purity: 98%
CAS No.: 7449-74-3
Molecular Formula: C6H15NOSi
Formula Weight: 145.28
Flash Point: 48°(118°F)
Refractive Index: 1.4390
Boiling Point: 48-49°/11mm
Beilstein Reference: 506975
UN No.: UN1993
MDL No.: MFCD00040758
Formula:
C₆H₁₅NOSi MW: 145,28 g/mol Boiling Pt: 48…49 °C (11 mmHg) Density: 0,906 Flash Pt: 48 °C (118 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00040758 CAS Number: 7449-74-3 EINECS: 231-217-0 UN: 1993 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...