2-Chloroethyl-p-tolyl sulphone 98%
Supplier: Thermo Scientific
L05256.03
L05256.06
ALFAL05256.03EA
0
SGD
ALFAL05256.03
ALFAL05256.06
2-Chloroethyl-p-tolyl sulphone 98%
2-Chloroethyl-p-tolyl sulphone
Purity: 98%
CAS No.: 22381-53-9
Molecular Formula: C9H11ClO2S
Formula Weight: 218.70
Melting Point: 77-81°
Beilstein Reference: 1107627
MDL No.: MFCD00051607
Formula:
C₉H₁₁ClO₂S MW: 218,70 g/mol Melting Pt: 77…81 °C Storage Temperature: Ambient |
MDL Number:
MFCD00051607 CAS Number: 22381-53-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...