1-Dimethylamino-2-pentyne 98%

Supplier: Thermo Scientific
Danger

L05662.06 L05662.14
ALFAL05662.06EA 0 SGD
ALFAL05662.06 ALFAL05662.14
1-Dimethylamino-2-pentyne 98%
1-Dimethylamino-2-pentyne

Purity: 98% CAS No.: 7383-77-9 Molecular Formula: C7H13N Formula Weight: 111.19 Refractive Index: 1.4360 Beilstein Reference: 1738713 UN No.: UN2734 MDL No.: MFCD00041631

Formula: C₇H₁₃N
MW: 111,19 g/mol
Density: 0,812
Storage Temperature: Ambient
MDL Number: MFCD00041631
CAS Number: 7383-77-9
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR