3-(2-Furyl)-2-phenylpropenal 97%

Supplier: Thermo Scientific

L06220.06 L06220.14
ALFAL06220.06EA 70.3 SGD
ALFAL06220.06 ALFAL06220.14
3-(2-Furyl)-2-phenylpropenal 97%
3-(2-Furyl)-2-phenylpropenal

Purity: 97% CAS No.: 57568-60-2 Molecular Formula: C13H10O2 Formula Weight: 198.22 Flash Point: 110°(230°F) Melting Point: 57-60° Beilstein Reference: 5923 MDL No.: MFCD00036566

Formula: C₁₃H₁₀O₂
MW: 198,22 g/mol
Melting Pt: 57…60 °C
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00036566
CAS Number: 57568-60-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR