Ethyl-4-bromo-3-ethoxy-2-butenoate 90% stabilised, tech.
Supplier: Thermo Scientific
|
Danger
|
Stabilization:
Stabilised with sodium hydroxide
L06849.03
L06849.06
ALFAL06849.03EA
65.8
SGD
ALFAL06849.03
ALFAL06849.06
Ethyl-4-bromo-3-ethoxy-2-butenoate 90% stabilised, tech.
Ethyl-4-bromo-3-ethoxy-2-butenoate
Stab. with sodium carbonate
Purity: 90%
CAS No.: 1116-50-3
Molecular Formula: C8H13BrO3
Formula Weight: 237.10
Storage Temperature: 2°C to 8°C
Refractive Index: 1.4960
Odour: Pungent
Boiling Point: 88-90°/1mm
Beilstein Reference: 2042896
UN No.: UN3265
MDL No.: MFCD00051757
Formula:
C₈H₁₃BrO₃ MW: 237,10 g/mol Boiling Pt: 88…90 °C (1 mmHg) Storage Temperature: Refrigerator |
MDL Number:
MFCD00051757 CAS Number: 1116-50-3 UN: 3265 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...